Learn More
Eisco™ Cone Clutch Drive (Single-Sided) Mechanical Demonstration Model
SCI_ED
Catalog No. S27668 Shop All Eisco Products
Change view
Click to view available options
Cone Clutch Drive (Single-Sided) Mechanical Demonstration Model
Catalog No.
S27668 Cone Clutch Drive (Single-Sided) Mechanical Demonstration Model
1 options
Catalog No. S27668 Supplier Eisco™ Supplier No. PH0366TM26
Add to Cart
Add to Cart
The Cone Clutch Drive is an interactive visual demonstration of a mechanical power transmission system that uses cone-shaped friction surfaces to transfer torque between rotating shafts.
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how a clutch works in a drive system.
  • Hand crank and a push button to engage clutch on the opposing side.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Product Type Cone Clutch Drive (Single-Sided) Mechanical Demonstration Model