Description
- Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
- Cover-only option allows you to use your own ratchet straps or add backup covers to your tower skirt cover sets
- Perfect for manways that need to be locked out or secured, but little or no flange is present
- Make it last – unlike wasteful warning tape, these covers are designed to be reused for multiple projects
Specifications
Specifications
| Product Type | Confined Space Cover |
| Color | Black/Red |
| Material | Nylon |
| Dimensions (L x W) | 40 x 40 in. |
| Displayed Text | Permit required. No unauthorized entry. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.