Learn More
Eisco™ Erlenmeyer Flasks - Wide Neck
SCI_ED
Catalog No. S23909 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S23909 500 mL
S88914 50 mL
S27997 100 mL
S15471 250 mL
S23910 1000 mL
S15472 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S23909 Supplier Eisco™ Supplier No. CH0426C
Add to Cart
Add to Cart
High quality borosilicate 3.3 glass Erlenmeyer flasks with wide neck design.
  • Conical Erlenmeyer filtering flask with graduations
  • Made of high quality borosilicate 3.3 glass
  • Wide neck design
  • Conforms to DIN ISO 1773 standard
  • Autoclavable
Warranty and Services
1 year limited manufacturer warranty.
Specifications
Certifications/Compliance DIN ISO 24450 standard
Capacity (Metric) 500 mL
Material Borosilicate Glass
Neck Style Wide
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Wide
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment Untreated