Learn More
Thermo Scientific™ Connect Edge Adapter Kit for InstrumentConnect Remote Monitoring with OPC UA™
Catalog No. 01672511 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Remote Monitoring Device Adapter
Remote Monitoring Sensor Adapter
Wireless Monitoring Kit
For Use With (Equipment):
CO2, BSC and ULT, See thermofisher.com/edgecompatible for model details
Connect Edge Hardware, CO2/ULT Equipment and Devices
Connect Edge Hardware, Refrigerators/Freezers and Devices
Connect Edge Sensors
Connect Edge, CO2 Incubators and Devices
Connect Edge, Laboratory Equipment and Devices
Incubators, Environmental Chambers, See thermofisher.com/edgecompatible for model details
Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details
Includes:
Connect Edge Ethernet Device Adapter Kit with RS232 connection, power cable 2-3 m, mounting bracket.
Connect Edge Ethernet Device Adapter Kit with RS485 connection, power cable 2-3 m, mounting bracket.
Connect Edge Ethernet Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), mounting bracket, no power supply
Connect Edge Ethernet Device Adapter Kit with USB connection, power cable 2-3 m, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with RS485, (optional Ethernet when combined with EDGEKTADAPT99)
Connect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
Connect Edge Wi-Fi Device Adapter Kit with USB connection, power cable and 2-3 m (depending on kit), power supply, mounting bracket.
Ethernet Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3M Cable
Optional PoE Converter for Wi-Fi/Ethernet Adapters
Wi-FI (optional Ethernet when combined with EDGEKTADAPT99)
Wi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m Cable, Power Supply and Country Converters. Convert to Ethernet with EDGEKTADPT99 (no PoE)
Wi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m power cable, powers supply and country converters.
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Includes
01-672-511 Remote Monitoring Device Adapter Connect Edge Hardware, CO2/ULT Equipment and Devices Connect Edge Wi-Fi Device Adapter Kit with RS485, (optional Ethernet when combined with EDGEKTADAPT99)
11-731-054 Wireless Monitoring Kit Connect Edge Sensors Wi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m Cable, Power Supply and Country Converters. Convert to Ethernet with EDGEKTADPT99 (no PoE)
01-672-498 Remote Monitoring Device Adapter Connect Edge Hardware, Refrigerators/Freezers and Devices Wi-FI (optional Ethernet when combined with EDGEKTADAPT99)
11-731-074 Wireless Monitoring Kit CO2, BSC and ULT, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply mounting bracket.
11-731-075 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
01-672-503 Remote Monitoring Device Adapter Connect Edge, CO2 Incubators and Devices Wi-FI (optional Ethernet when combined with EDGEKTADAPT99)
11-731-076 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
01-672-507 Remote Monitoring Device Adapter Connect Edge, Laboratory Equipment and Devices Optional PoE Converter for Wi-Fi/Ethernet Adapters
11-731-053 Wireless Monitoring Kit Incubators, Environmental Chambers, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply, mounting bracket.
11-731-035 Wireless Monitoring Kit CO2, BSC and ULT, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
11-731-036 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
11-731-055 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
11-731-078 Wireless Monitoring Kit Incubators, Environmental Chambers, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with USB connection, power cable and 2-3 m (depending on kit), power supply, mounting bracket.
11-731-050 Wireless Monitoring Kit Connect Edge Sensors Wi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m power cable, powers supply and country converters.
13-245-175 Wireless Monitoring Kit CO2, BSC and ULT, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
13-245-176 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
13-245-177 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
13-245-178 Wireless Monitoring Kit Incubators, Environmental Chambers, See thermofisher.com/edgecompatible for model details Connect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
13-245-179 Wireless Monitoring Kit CO2, BSC and ULT, See thermofisher.com/edgecompatible for model details Connect Edge Ethernet Device Adapter Kit with RS485 connection, power cable 2-3 m, mounting bracket.
13-245-180 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Ethernet Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), mounting bracket, no power supply
13-245-181 Wireless Monitoring Kit Refrigerator/Freezer, See thermofisher.com/edgecompatible for model details Connect Edge Ethernet Device Adapter Kit with RS232 connection, power cable 2-3 m, mounting bracket.
13-245-182 Wireless Monitoring Kit Incubators, Environmental Chambers, See thermofisher.com/edgecompatible for model details Connect Edge Ethernet Device Adapter Kit with USB connection, power cable 2-3 m, mounting bracket.
13-245-183 Remote Monitoring Sensor Adapter Connect Edge Sensors Ethernet Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3M Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 01-672-511 Supplier Thermo Scientific™ Supplier No. EDGEKTADPT01
Add to Cart
Add to Cart
Connect Edge Wi-Fi/Ethernet device adapters are for use with InstrumentConnect 24/7 remote monitoring system. Device adapters plug into Thermo Fisher laboratory equipment to extract and transmit telemetry to the Connect Edge gateway via Wi-Fi or Ethernet. Configuration needed varies by equipment type.
Connect Edge Wi-Fi/Ethernet device adapters are part of the InstrumentConnect 24/7 remote monitoring system. Device adapters include RS485, TTL, RS232 and USB configurations to plug into your Thermo Fisher laboratory equipment. Device adapters extract and transmit telemetry to the Connect Edge gateway via Wi-Fi or Ethernet.
Thermo Scientific™ InstrumentConnect is professional-grade remote monitoring system for monitoring laboratory equipment and other devices to help protect samples and increase asset management efficiencies. The InstrumentConnect system utilizes Connect Edge adapters and gateways to send data to the InstrumentConnect cloud or to your own premise-based system (LIMS, BMS, SCADA) using OPC-UA™ protocol. Connect Edge hardware is compatible with 700+ Thermo Fisher Scientific laboratory equipment makes and models.
Thermo Fisher Scientific Equipment Telemetry
  • For cold storage equipment, InstrumentConnect collects ambient and cabinet temperature data recorded by resistance temperature detectors (RTDs) built into the unit; the remote monitoring system also reports alarms and events, as well as other system-based diagnostic data points to the InstrumentConnect application.
  • For CO2 incubators and environmental chambers, InstrumentConnect collects chamber temperatures, gas levels, alarms and alerts received via the incubator and environmental chamber data output connection.
System Hardware
  • Device Adapter-Palm-size adapters collect and transmit data from lab equipment/devices to gateways using Ethernet (WAN) connectivity or Wi-Fi.
  • Connect Edge Gateway-Receives data from device adapters to send to InstrumentConnect (cloud) or customer-premise system/cloud via OPC-UA using Ethernet (WAN) connectivity or Wi-Fi.
System Software
Choose from these available InstrumentConnect packages.
InstrumentConnect Monitor (Cloud):
  • Monitor onboard telemetry data and events**
  • Custom alarm settings
  • View live device status 24/7 via Mobile and Cloud app
  • Peace of mind with industry-leading security protocols
  • 25GB included data storage for documents
  • Unlimited device telemetry storage
  • Notifications via mobile application, e-mail and desktop application
InstrumentConnect Integrate (OPC-UA to Cloud or Prem):
  • Utilize the Connect Edge system to collect and transmit data to your on-premise system or cloud using Open Platform Communications Unified Platform. The Connect Edge gateway can be enabled as an OPC server for 10 or more devices.
Network
You can connect 10 device adapters to each Connect Edge gateway. Gateways can accommodate greater than 10 devices, however this is determined by reviewing CPU usage after you have reached this initial quantity after initial set up. Deciding which topology is applicable to your setting will determine the number of gateways you will need.
Connect Edge system has 3 main topologies
  • Direct Wi-Fi-Device adapters are configured to transmit data directly to your Connect Edge gateway (10 max per gateway). Your device adapters must be in proximity (50-150 feet) to the gateway.
  • Single/Dual Network-Device adapters are configured to connect to your local network via Ethernet or Wi-Fi and transmit data to the Connect Edge gateway, also on the same network. This topology is ideal if devices to be monitored are not within proximity to each other or a gateway.
Building your system
  • Determine type (Ethernet-only/WiFi+Ethernet) and quantity of gateway(s) needed based on network topology preference. The Configuration Planning Guide can be used for reference, if needed.
  • Select compatible adapter for your lab equipment
  • Purchase an InstrumentConnect Monitor or Integrate subscription for each device
  • Determine whether self-install or professional-install is desired (where available)
System hardware and software is available for purchase in packages or a-la-carte. A built-in gateway discount is included with all packages. Additional savings with 3 and 5-year packages.
Installation
Customer (self) installation
Installation services sold separately, if desired (US, Europe, Great Britain, inquire with specialist for other regional availability).
Availability
InstrumentConnect is available in the following countries and configurations. For the most updated list, please ask your specialist.
Wi-Fi or Ethernet gateway/adapter: Europe, UK, North America, Australia, New Zealand, Puerto Rico
Ethernet-only gateway/Wi-Fi or Ethernet adapter: Hong Kong
Ethernet-only gateway and adapter: Brazil, Bahrain, Oman, UAE
TRUSTED_SUSTAINABILITY
Specifications
Type Remote Monitoring Device Adapter
Display Type No Display
Alarm Types None
Software Type InstrumentConnect
Includes Connect Edge Wi-Fi Device Adapter Kit with RS485, (optional Ethernet when combined with EDGEKTADAPT99)
Interface RS485 at the device
IP Rating IP30
Quantity 1 Ea.
Warranty 1 Year Warranty
For Use With (Application) InstrumentConnect Remote Monitoring System, Laboratory Equipment and Devices
For Use With (Equipment) Connect Edge Hardware, CO2/ULT Equipment and Devices
Electrical Requirements AC wall adapter (provided)