Learn More
Connecticut Clean Room™ PolyTack Mats
Catalog No. 19062728
Change view
Click to view available options
Color:
Blue
White
Dimensions (L x W):
18 x 36 in.
26 x 45 in.
36 x 36 in.
36 x 45 in.
36 x 60 in.
36 x 72 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
19-062728 Blue 36 x 72 in.
19-808-714 Blue 36 x 36 in.
19-086599 White 36 x 36 in.
19-808-713 Blue 18 x 36 in.
19-064223 Blue 36 x 45 in.
19-808-715 Blue 26 x 45 in.
19-085148 Blue 36 x 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-062728 Supplier Connecticut Clean Room™ Supplier No. P108B
Add to Cart
Add to Cart
Removes foot-borne particulate before entering the cleanroom
  • Made from an incinerable polyethylene film
  • Seamless one-piece construction
  • Numbered tabs provide easy identification and sheet separation
  • Easy mobility for carts and wheeled machinery
  • Polyethylene film contains a special anti-microbial agent providing long-lasting protection against bacterial growth
Specifications
Dimensions (L x W) 36 x 72 in.
Material Polyethylene
Color Blue