Learn More
Mettler Toledo™ Connector Syringe/Filling Tube
Catalog No. 01915183 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
Densito and DensitoPro Handheld Density Meters
Catalog No. For Use With (Equipment)
01-915-183 Densito and DensitoPro Handheld Density Meters
1 options
Catalog No. 01-915-183 Supplier Mettler Toledo™ Supplier No. 30330853
Add to Cart
Add to Cart
Mettler Toledo Connector Syringe/Filling Tube are ideal for use with viscous samples
  • When working with viscous samples, this connector acts as an adapter between the Density2Go handheld meter and the syringe, making your work easier
TRUSTED_SUSTAINABILITY
Specifications
Languages Chinese, English (GB), English, French, German, Italian, Korean, Polish, Brazilian Portuguese, Russian, Spanish
Description Connector Syringe/Filling Tube, Densito
For Use With (Equipment) Densito and DensitoPro Handheld Density Meters
For Use With (Application) Use with Densito and DensitoPro Handheld Density Meters