Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin 26 Monoclonal Antibody (CX-1E8), Invitrogen™

Catalog No. PI335800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI335800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI335800 Supplier Thermo Scientific Supplier No. 335800

Mouse Monoclonal Antibody

33-5800 was used in the immunofluorescence and western blot analysis to successfully detect Connexin26 in mouse liver sections and homogenate, respectively.

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variables (EKV) is an autosomal dominant genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 26
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone CX-1E8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide, pH 7.4
Gene Gjb2
Gene Accession No. P21994, Q00977
Gene Alias AI325222, Cnx26, connexin 26, connexin-26, CX26, CXN-26, DFNA3, DFNA3A, DFNB1, DFNB1A, Gap junction beta-2 protein, gap junction channel protein connexin 26, gap junction membrane channel protein beta 2, gap junction protein beta 2, gap junction protein, beta 2, gap junction protein, beta 2, 26kDa, GJB2, Gjb-2, HID, KID, mutant gap junction protein beta 2, NSRD1, PPK
Gene Symbols Gjb2
Host Species Mouse
Immunogen A 13 amino acid synthetic peptide derived from the C-terminus of the mouse Connexin 26 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14619, 394266
Target Species Mouse, Rat
Content And Storage -20°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.