Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 30.2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 407400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-740-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-740-0 Supplier Invitrogen™ Supplier No. 407400
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

40-7400 was used in the western blot analysis to successfully detect Connexin 30.2 in HeLa cells transfected with Cx30.2 and in immunofluorescence (IHC) analysis to successfully detect Connexin 30.2 in mouse heart sections.

Gap junctions are responsible for transport of ions and metabolites between adjacent cells. These structures are made of two hemichannels, each formed by six connexin molecules. Connexin 30, coded by Cjb6 gene, is a member of the connexin family and is highly expressed in brain and skin. Immunohistochemically connexin 30 was localized in astrocytes, at gap junctions between these cells and on the astrocyte side of gap junctions between astrocytes and oligodendrocytes. Co-localization with connexin 43 was also observed in this study. Cytoskeleton, especially actin filaments, are important components in the processes of assembly, trafficking and stabilization of connexin 30 gap junctions. Knock-out mice studies have also demonstrated that connexin 30 deficiency impairs renal tubular ATP release and pressure natriuresis highlighting the importance of this molecule in kidney. Further, in vitro studies have also demonstrated that connexin 30 overexpression enhances cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 30.2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJD3
Gene Accession No. Q8N144, Q91YD1
Gene Alias connexin 30.2; connexin 31.9; Connexin-30.2; Connexin-31.9; Cx30.2; Cx31.9; gap junction alpha-11 protein; Gap junction chi-1 protein; gap junction delta-3 protein; gap junction membrane channel protein chi 1; gap junction protein delta 3; gap junction protein, delta 3; gap junction protein, delta 3, 31.9kDa; Gja11; GJC1; GJD3
Gene Symbols GJD3
Host Species Rabbit
Immunogen Synthetic peptide derived from the c-term region of the mouse connexin 30.2 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125111, 353155
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.