Learn More
Description
Connexin 37 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | Connexin 37 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | GJA4 |
| Gene Accession No. | P28235, P35212, Q03190 |
| Gene Alias | AU020209; AW558810; Cnx37; connexin 37; connexin-37; CX37; CXN37; Cxn-37; gap junction alpha-4 protein; gap junction membrane channel protein alpha 4; Gap junction membrane channel, protein alpha 4 (connexin 37); gap junction protein alpha 4; gap junction protein, alpha 4; gap junction protein, alpha 4, 37kDa; GJA4; Gja-4 |
| Gene Symbols | GJA4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
