Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin-37 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985878
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5878 Supplier Bioss Supplier No. BS4067R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene have been associated with atherosclerosis and a higher risk of myocardial infarction.

Specifications

Antigen Connexin-37
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GJA4
Gene Accession No. P35212
Gene Alias AU020209; AW558810; Cnx37; connexin 37; Connexin-37; Cx37; CXN37; Cxn-37; Gap junction alpha-4 protein; gap junction membrane channel protein alpha 4; Gap junction membrane channel, protein alpha 4 (connexin 37); gap junction protein alpha 4; gap junction protein, alpha 4; gap junction protein, alpha 4, 37kDa; GJA4; Gja-4
Gene Symbols GJA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Connexin 37.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2701
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.