Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 47 Monoclonal Antibody (4A11A2)
GREENER_CHOICE

Catalog No. 374500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-450-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-450-0 Supplier Invitrogen™ Supplier No. 374500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Connexin 47 Monoclonal Antibody for Western Blot, IHC, IP

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 47
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4A11A2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJC2
Gene Accession No. Q8BQU6
Gene Alias B230382L12Rik; connexin 47 type A; connexin 47 type C; connexin 47 type D; connexin-46.6; Connexin-47; CX46.6; Cx47; CXG2; gap junction alpha-12 protein; Gap junction gamma-2 protein; gap junction membrane channel protein alpha 12; gap junction protein gamma 2; gap junction protein, alpha 12; gap junction protein, chi 2; gap junction protein, gamma 2; gap junction protein, gamma 2, 47kDa; GJA12; Gjc2; HLD2; LMPH1C; PMLDAR; SPG44
Gene Symbols GJC2
Host Species Mouse
Immunogen Synthetic peptide derived from the C-terminal region of the mouse Connexin 47 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118454
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.