Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Connexin 50 Monoclonal Antibody (C6)
GREENER_CHOICE

Catalog No. 334300
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-430-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-430-0 Supplier Invitrogen™ Supplier No. 334300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Connexin 50 Monoclonal Antibody for ELISA, ICC/IF, IP, Western Blot

This gene encodes a transmembrane connexin protein that is necessary for lens growth and maturation of lens fiber cells. The encoded protein is a component of gap junction channels and functions in a calcium and pH-dependent manner. Mutations in this gene have been associated with zonular pulverulent cataracts, nuclear progressive cataracts, and cataract-microcornea syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 50
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJA8
Gene Accession No. P28236, P48165, P55917, Q8K4Q9
Gene Alias Aey5; alpha 8 connexin; CAE; CAE1; cell surface glycoprotein; Cnx50; connexin 50; connexin-49; Connexin-50; CTRCT1; cx49; CX50; CZP1; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha 8; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50kDa; gap junction protein, alpha 8; gap junction protein, alpha 8, 50kDa; GJA8; Lens fiber protein MP70; lens intrinsic membrane protein MP70; lens opacity 10; Lop10; MP38; MP64; MP70
Gene Symbols GJA8
Host Species Mouse
Immunogen This antibody was prepared against urea/alkali stripped sheep lens membranes.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100170231, 14616, 2703, 29601
Target Species Human, Mouse, Rat, Sheep
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.