Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Connexin Antibody Sample Pack

Catalog No. 900500
Encompass_Preferred
Change view
Click to view available options
Quantity:
90 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-050-0 90 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-050-0 Supplier Invitrogen Supplier No. 900500
Add to Cart
Add to Cart

Mouse Polyclonal, Monoclonal Antibody

Specificity: Connexin 26, 32, 43

Reactivity: Human, Mouse, Rat

Applications: See product data sheet for recommended Usage for each antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal/Polyclonal
Clone Sampler Pack
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 90 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2697
Target Species Human, Mouse, Rat
Content And Storage Store in freezer (-5 to -30°C).
Reactivity Human, Mouse, Rat
Shipping Condition Wet Ice
Reactivity abrv Rt, Hu, Ms
Target Gene Connexin
Validated Application abrv WB, IHC (Frozen), ELISA
Product Line GeneChip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.