Learn More
Therapak™ ConstanTemp™ Specimen Insulator for Lock Boxes
Catalog No. p-4121051 Shop All Therapak Corporation Products
Change view
Click to view available options
Color:
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-130-218 Blue
22-131-454 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-130-218 Supplier Therapak™ Supplier No. 36525
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
An ideal source of protection for maintaining specimen integrity prior to courier pick up in lock boxes or in courier transit
Features a fabric-based construction and Volara insulation combined with specially formulated gel that holds temperature for extended periods of time. For frozen specimens, place ConstanTemp in the freezer overnight. When specimen is frozen, insert it into the pouch and place in the lock box for transport. It will stay frozen over a period of 19 hours at an ambient temperature of 22°C. For refrigerated specimens, place insulator into refrigerator overnight or until it reaches 2°C. It will maintain 2–8°C in either a hot or cold environment for hours. For room temperature or ambient specimens, leave insulator on the counter in a 20-25°C room and place specimen inside. It will maintain this temperature for hours.
  • Maintains ambient, refrigerated and frozen specimen integrity
  • Custom printing and fabric color options available
  • Blue color (also available in red and white; contact your Fisher HealthCare representative)
Specifications
Color Blue