Learn More
Ampco Safety Tools Construction Wrench (Metric)
SDP
Catalog No. 19174644 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric) Opening:
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
23 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
32 mm
33 mm
34 mm
35 mm
36 mm
37 mm
38 mm
41 mm
42 mm
46 mm
47 mm
50 mm
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Opening
19-174-644 22 mm
19-174-638 17 mm
19-174-639 18 mm
19-174-640 19 mm
19-174-642 20 mm
19-174-643 21 mm
19-174-645 23 mm
19-174-646 24 mm
19-174-647 25 mm
19-174-648 26 mm
19-174-649 27 mm
19-174-650 28 mm
19-174-651 29 mm
19-174-653 30 mm
19-174-654 32 mm
19-174-655 33 mm
19-174-656 34 mm
19-174-657 35 mm
19-174-658 36 mm
19-174-659 37 mm
19-174-660 38 mm
19-174-662 41 mm
19-174-663 42 mm
19-174-664 46 mm
19-174-665 47 mm
19-174-667 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-644 Supplier Ampco Safety Tools Supplier No. W2222
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Construction Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Head Style Construction, Offset Type with Pin
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Length (Metric) Opening 22 mm
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.