Learn More
Chemglass Life Sciences Digital Thermoregulator ONLY, with Built In Timer
Supplier: Chemglass Life Sciences CG1995V20
Digital Thermoregulator ONLY, with Built In Timer. Digital thermoregulator is equipped with innovative Fuzzy Logic technology that provides accurate set point +/- 0.5 C. Digital count down timer shuts off heating- adjustable from 0 to 24:59 (h:min). Innovative design incorporated a clamp for mounting on the vertical support rod.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.