Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Contactin 2 Monoclonal Antibody (8B9F4G1)
GREENER_CHOICE

Catalog No. PIMA529178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529178 Supplier Invitrogen™ Supplier No. MA529178
  • $410.00
    $339.65 / Each of 1

    Save $70.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human CNTN2/TAG-1.

Contactin 2 (CNTN2, transiently-expressed axonal glycoprotein, AXT, TAX, TAX1, TAG-1, axonin 1) is a neuronal cell adhesion molecule (CAM) that influences the formation of axon connections in the developing nervous system. Contactin 2 is a member of the immunoglobulin superfamily (IgSF) and contains a glycosylphosphatidylinositol-anchor, six immunoglobulin (Ig)-like and four Fibronectin type III (FNIII)-like domains. Contactin 2 is expressed predominantly during neural development on the cell membrane of axons in nerve fiber tracts in order to guide commissural axons without promoting their growth. Contactin 2 binds with NgCAM in the plane of the same membrane (cis-binding). The Contactin 2 heterophilic (Contactin 2/NgCAM and Contactin 2/NrCAM) binding sites are localized to the first four Ig domains. The Contactin 2 homophilic (Contactin 2/Contactin 2) binding site is localized to the FNIII domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Contactin 2
Applications ELISA
Classification Monoclonal
Clone 8B9F4G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CNTN2
Gene Accession No. Q02246
Gene Alias Axonal glycoprotein TAG-1; axonin; Axonin 1; Axonin1; axonin-1; axonin-1 cell adhesion molecule; AXT; Cntn2; contactin 2; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin2; contactin-2; D130012K04Rik; DKFZp781D102; FAME5; MGC157722; TAG1; TAG-1; TAG-564; Tax; TAX1; TAX-1; transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein
Gene Symbols CNTN2
Host Species Mouse
Immunogen Human cell-derived rhCNTN2 (Met1-Asn1012).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6900
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.