Learn More
Invitrogen™ Contactin 6 Polyclonal Antibody

Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for increased stability, and at 4 °C with an appropriate antibacterial agent. Avoid repetitive freeze/thaw cycles. Glycerol (1:1) may be added for an additional stability.
Specifications
Specifications
| Antigen | Contactin 6 |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with no preservative |
| Gene | CNTN6 |
| Gene Accession No. | P97528, Q9JMB8, Q9UQ52 |
| Gene Alias | CNTN6; contactin 6; contactin-6; hNB-3; mNB-3; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-3 |
| Gene Symbols | CNTN6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.