Learn More
Thomas Scientific Flip-Top Waste Container
SDP
Catalog No. 501461087
Encompass
IBI’s flip-top waste containers are designed for the safe disposal of contaminated pipette tips, microcentrifuge tubes, mini columns, as well as any other small items that require containment. The FL-60 and FL-150’s lid has a Flip-Top cap with a 2cm diameter entry hole under it for disposal without having to remove the entire lid, thus limiting exposure to the user. The full lid also lifts off for disposal of larger items and removal when full. The containers are designed to accept NRC approved bags and are constructed of heavy-duty 1/2” cast acrylic for years of use and maximum protection.
Catalog No. 50-146-1087 Supplier Thomas Scientific Supplier No. 1226L55
IBI’s flip-top waste containers are designed for the safe disposal of contaminated pipette tips, microcentrifuge tubes, mini columns, as well as any other small items that require containment. The FL-60 and FL-150’s lid has a Flip-Top cap with a 2cm diameter entry hole under it for disposal without having to remove the entire lid, thus limiting exposure to the user. The full lid also lifts off for disposal of larger items and removal when full. The containers are designed to accept NRC approved bags and are constructed of heavy-duty 1/2” cast acrylic for years of use and maximum protection.