Learn More
Med Vet International Triangular intake / Output Container, Graduated, Clear, 10/Pack
Supplier:  Med Vet International DYND80416H
Encompass_Preferred
Triangular Graduates: 32-oz. plastic triangular container is graduated in mL and ounces for measuring intake or output accurately. Triangular style allows ease of measuring, visualization of contents, and comfortable handling. Features frosted block to include important patient information. Etched Graduations Clear, Rigid.
Catalog No. 50-282-9858
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.