Learn More
Contec™ Bucket Wringers and Frames
BIOPHARMAGREENER_CHOICE
Catalog No. 19168528 Shop All Contec Inc. Products
Change view
Click to view available options
Product Type:
Curved Mop Wringer
Flat Mop Wringer
Heavy Duty Downpress Mop Wringer
Slinger Frame and Insert
For Use With (Equipment):
Contec EasyCurve Mop
Contec EasyCurve Mops, 6.5 gal. Bucket Only
Contec Edgeless VertiKlean MAX or QuickTask Mops, 5 gal. or 6.5 gal. Bucket
Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket
Contec VertiKlean MAX or QuickTask Flat Mops, 6.5 gal. Bucket
VertiKlean MAX and QuickTask Mops
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
19-168-528 Flat Mop Wringer VertiKlean MAX and QuickTask Mops
19-033-705 - Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket
19-168-797 - Contec VertiKlean MAX or QuickTask Flat Mops, 6.5 gal. Bucket
19-168-529 Curved Mop Wringer VertiKlean MAX and QuickTask Mops
19-200-014 - Contec EasyCurve Mop
19-159-255 Heavy Duty Downpress Mop Wringer Contec Edgeless VertiKlean MAX or QuickTask Mops, 5 gal. or 6.5 gal. Bucket
19-033-706 Slinger Frame and Insert Contec EasyCurve Mops, 6.5 gal. Bucket Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-168-528 Supplier Contec™ Supplier No. 2691
Add to Cart
Add to Cart
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
  • Choose from downpress, flat or curved styles
  • Available in polypropylene or stainless steel
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Select wringer that complements mop
Curved stainless-steel sieve 19-200-014 for EasyCurve™ mop and flat stainless-steel sieve 19-033-715 for flat mops such as VertiKlean™ MAX and QuickTask. Sieves require stainless-steel frame 19-033-716 for 5 gal. (20 L) buckets and 19-033-717 for 6.5 gal. (25 L) buckets.
Specifications
Product Type Flat Mop Wringer
Material Stainless Steel
Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Application) Ideal for cleanrooms and controlled environments
For Use With (Equipment) VertiKlean MAX and QuickTask Mops
Sterility Non-sterile