Description
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
- Choose from downpress, flat or curved styles
- Available in polypropylene or stainless steel
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- Select wringer that complements mop
Specifications
Specifications
| Product Type | Curved Mop Wringer |
| Material | Stainless Steel |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| For Use With (Application) | Ideal for cleanrooms and controlled environments |
| For Use With (Equipment) | VertiKlean MAX and QuickTask Mops |
| Sterility | Non-sterile |