Learn More
Contec™ 6.5 Gallon Bucket System
BIOPHARMA
Catalog No. 19200007 Shop All Contec Inc. Products
Change view
Click to view available options
Capacity (English):
2 x 6.5 gal. Buckets
Catalog No. Capacity (English)
19-200-007 2 x 6.5 gal. Buckets
1 options
Catalog No. 19-200-007 Supplier Contec™ Supplier No. 2772KIT
Includes
Two polypropylene 6.5 gallon (25L) buckets (red, gray) and a stainless-steel cart with standard or heavy-duty casters.
Compact design is ideal for smaller cleanrooms
  • Compact design is ideal for smaller cleanrooms
  • Contec's Bucket System 2772-KIT is a fully autoclavable stainless steel rolling cart with two 6.5 gallon (25 L) polypropylene buckets
  • The lightweight, yet substantial stainless steel cart with heavy-duty casters is easy to maneuver around any size cleanroom
  • The quiet, heavy-duty wheels feature stainless steel frames and locking brakes
  • The red and gray double bucket system allows for differentiation between cleaning and rinsing solutions
  • The autoclavable polypropylene buckets are compatible with two stainless steel sieves (sold separately)
  • 2768 is a curved sieve that is used with our EasyCurve™ mop. 2767 is a flat sieve for Contec VertiKlean MAX and QuickTask flat mops.
  • Both sieves are used with the stainless steel 2766 frame designed for our 6.5 gallon (25 L) buckets
Specifications
Product Type Rolling Cart Double Bucket System
Material Polypropylene
Autoclavable Autoclavable
Capacity (English) 2 x 6.5 gal. Buckets
For Use With (Application) Compact Design is ideal for Smaller Cleanrooms
Includes Two 6.5 gal. (25 L) Buckets (Gray, Red), Stainless Steel Cart with Heavy Duty Casters