Learn More
Contec™ VertiKlean™ MAX™ Mop Heads
BIOPHARMAGREENER_CHOICE
Catalog No. 19168810 Shop All Contec Inc. Products
Change view
Click to view available options
Dimensions:
21 x 11 x 2 cm
24 x 13 x 1 cm
32 x 11 x 2 cm
38 x 11 x 2 cm
38 x 13 x 1 cm
46 x 13 x 1 cm
Sterility:
Non-sterile
Sterile
Material:
Hydrophilic Urethane Foam
Polyester, Foam
Ribbed Polyester Microfiber, Foam
Ribbed Polyester Microfiber, Hydrophilic Urethane Foam
Head Type:
Non-sealed Edge
Ribbed Sealed Edge
Sealed Edge
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions Sterility Material Head Type
19-168-810 38 x 13 x 1 cm Sterile Ribbed Polyester Microfiber, Foam Ribbed Sealed Edge
19-159-236 21 x 11 x 2 cm Non-sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-159-237 21 x 11 x 2 cm Sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-159-238 32 x 11 x 2 cm Non-sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-159-239 32 x 11 x 2 cm Sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-159-240 38 x 11 x 2 cm Non-sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-159-241 38 x 11 x 2 cm Sterile Ribbed Polyester Microfiber, Foam Non-sealed Edge
19-168-811 38 x 13 x 1 cm Non-sterile Ribbed Polyester Microfiber, Foam Ribbed Sealed Edge
19-200-050 38 x 13 x 1 cm Sterile Ribbed Polyester Microfiber, Foam Sealed Edge
19-176-942 24 x 13 x 1 cm Non-sterile Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge
19-176-943 24 x 13 x 1 cm Sterile Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge
19-169-127 46 x 13 x 1 cm Non-sterile Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge
19-169-128 46 x 13 x 1 cm Sterile Ribbed Polyester Microfiber, Hydrophilic Urethane Foam Sealed Edge
19-991-017 38 x 13 x 1 cm Non-sterile Hydrophilic Urethane Foam Sealed Edge
19-165-63 21 x 11 x 2 cm Non-sterile Polyester, Foam Non-sealed Edge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-168-810 Supplier Contec™ Supplier No. VKSE0250
Add to Cart
Add to Cart
This item is not returnable. View return policy
Clean walls, ceilings and other hard-to-reach surfaces with ease. VertiKlean™ MAX™ Mop Heads easily snap onto frames and last longer than traditional mops.
  • Features a single layer of hydrophilic polyurethane foam laminated to textured polyester microfiber fabric
  • Backed by a semi-rigid thermoformed plate that easily snaps onto mop head frame
  • Designed for applying disinfectants to both horizontal and vertical surfaces
  • Mop heads are composed of a disposable absorbent sponge laminated to filament polyester knit fabric and backed with a thermoformed plate that easily snaps onto VertiKlean mop hardware
  • Now available with ribbed microfiber for better contamination removal
  • Pivoting mop head frame is ideal for cleaning walls, floors, and ceilings as well as hard-to-reach areas such as fume hoods, mini environments, and around light fixtures and heavy equipment
  • Sterile mop heads are validated sterile per AAMI ISO11137 guidelines
  • Autoclaving mop heads is not recommended due to thermoformed plate; however, all hardware is autoclavable
Order Info
Frames, handles and bucket wringer system sold separately.
Certifications
Validated sterile to a Sterility Assurance Level of 10-6 per AAMI ISO11137:2006 Guidelines. Certificate of sterility is available.
Specifications
Dimensions 38 x 13 x 1 cm
Color Code White
Product Type Mop Head
Head Type Ribbed Sealed Edge
Sterility Sterile
Autoclavable Not Autoclavable
Disposable Single-use
Color-coded No
Material Ribbed Polyester Microfiber, Foam