Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ VertiKlean™ Sox Mop Head Covers
BIOPHARMAGREENER_CHOICE

Catalog No. 19051332 Shop All Contec Inc. Products
Change view
Click to view available options
Size:
Large
Medium
Regular
For Use With (Equipment):
Large VertiKlean Mop Head
Medium VertiKlean Mop Head
Regular VertiKlean Mop Head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Equipment)
19-051-332 Large Large VertiKlean Mop Head
19-159-256 Medium Medium VertiKlean Mop Head
19-150-0701 Regular Regular VertiKlean Mop Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-051-332 Supplier Contec™ Supplier No. LVKSOX
Add to Cart
Add to Cart

Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.

  • Clean-processed polyester
  • Extends the life of VertiKlean mop heads

Specifications

Color Code White
Product Type Mop Cover
Sterility Non-sterile
Autoclavable Not Autoclavable
Size Large
Disposable Single-use
For Use With (Equipment) Large VertiKlean Mop Head
For Use With (Application) Removable cover increases durability of VertiKlean mop when used on abrasive surfaces
Color-coded No
Material Polyester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.