Learn More
LABSYSTEMS DIAGNOSTICS OY Control sets for Vitamin A/E assay
Supplier:  LABSYSTEMS DIAGNOSTICS OY 72500333
Encompass_Preferred
VAE-Control set (Low + High Control) is intended to be used in clinical laboratories for the quantitative determination of vitamin A and E in serum and plasma to detect deficiencies or hypervitaminosis. The controls are prepared using human serum and in a lyophilized form
Catalog No. NC2865509