Learn More
Mopec™ Control Slides, Special Stains, Acid Fast Bacteria
SDP
Catalog No. 22457680 Shop All Mopec Products
Change view
Click to view available options
Includes:
20 Unstained, 2 Stained Slides
98 Unstained, 2 Stained Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-457-680 98 Unstained, 2 Stained Slides
22-457-726 20 Unstained, 2 Stained Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-457-680 Supplier Mopec™ Supplier No. SC801100
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 20 contains 20 unstained slides and 2 stained slides
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Includes 98 Unstained, 2 Stained Slides
Product Type Control Slide