Learn More
Mopec™ Control Slides, Special Stains, Hepatitis B
SDP
Catalog No. 22457488 Shop All Mopec Products
Change view
Click to view available options
Includes:
10 Unstained, 1 Stained Slide
20 Unstained, 2 Stained Slides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-457-488 10 Unstained, 1 Stained Slide
22-457-574 20 Unstained, 2 Stained Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-457-488 Supplier Mopec™ Supplier No. SC81610
Add to Cart
Add to Cart
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 20 contains 20 unstained slides and 2 stained slides
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Includes 10 Unstained, 1 Stained Slide
Product Type Control Slide