Learn More
Mopec™ Control Slides, Special Stains, Periodic Acid Schiff with Digestion (PAS Diastase Stain (PAS-D))
Shop All Mopec Products
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Specifications
| Includes | 20 Unstained, 2 Stained Slides |
| Product Type | Control Slide |