Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Controlled Environment Coverall

Catalog No. 19165012G Shop All Fisherbrand Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-165-012G 4X-Large
19-165-012A Small
19-165-012B Medium
19-165-012C Large
19-165-012D X-Large
19-165-012E 2X-Large
19-165-012F 3X-Large
19-165-012H 5X-Large
19-165-012I 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-165-012G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fisherbrand microporous composite coveralls are designed for many non-hazardous applications and will keep the wearer clean and comfortable all day.

  • Microporous composite material
  • Strong and lightweight
  • Tapered collar
  • Zipper front
  • Elastic wrist, ankle and back
  • Strong stress-resistant serged seams

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Collar Type Tapered
Back Style Elastic Back
Latex Free Yes
No. of Pockets None
Sleeve Style Set-in Sleeve
For Use With (Application) Controlled and Critical Environments
Quantity 25/Cs.
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.