Learn More
GSC Go Science Crazy Convection of Gases Apparatus
SCI_ED
Catalog No. S13645 Shop All GSC Products
Click to view available options
Gas convection apparatus; Educational
Catalog No.
S13645 Gas convection apparatus; Educational
1 options
Intended for ages 13 and up.
Catalog No. S13645 Supplier GSC Go Science Crazy Supplier No. 3031
Add to Cart
Add to Cart
Intended for ages 13 and up.
Includes
Candle; Smoke paper; Instructions
Demonstrates how convection currents work. The GSC International Convection of Gases Apparatus provides a compact, effective visual demonstration for students in classroom settings. Included instructions ensure easy operation and lesson success.
  • Provides a visual demonstration that makes it easy for students to see how convection currents work
  • A candle is lit below one of the chimneys, while an incense stick is held above the other chimney; the convection current will then carry the smoke down and out of the other chimney to visually represent the convection current
  • The box has a removable glass front, two chimneys, and two chimney covers
  • Includes candle, instructions, and everything needed for the demonstration
  • Ideal for classroom settings
Specifications
For Use With (Application) Demonstrating convection currents
Dimensions (L x W x H) 8.5 x 3.25 x 3.5 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov