Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex Safety Products Cooling Neck Shade
Shop All Pyramex Safety Products Products
Click to view available options
Color:
Lime
Orange
Description
- Evaporative cooling material keeps users cool for hours
- Made from soft quilted hi-vis material for greater visibility
- Fits on the suspension of a hard hat with hook and loop closures
- To activate, soak in cool water for 2 to 5 minutes
- Reusable – just reactivate
Specifications
Specifications
| Product Type | Cooling Product |
| Color | Lime |
| For Use With (Equipment) | Hard Hat |
| Size | Universal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction