Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coomassie Brilliant Blue G-250, MP Biomedicals™

Catalog No. MP219034380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219034380 100 g
ICN19034305 5 g
ICN808274 10 g
ICN19034325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219034380 Supplier MP Biomedicals, Inc Supplier No. 0219034380
Add to Cart
Add to Cart

Useful dye for SDS gels as it readily stains proteins with minimal background color

  • It is a protein stain in electrophoresis.
  • It has been used in the Bradford dye-binding protein assay.
  • A mechanism for dye binding to protein has been proposed, based on measuring Coomassie Brilliant Blue G-250 (CBBG) absorbance spectra during titration of the dye reagent in the absence of protein and its response to different polyamino acids.
  • Brilliant Blue G has been used for determining critical micelle concentration of detergents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
MDL Number MFCD00078482

Specifications

Beilstein 5230822
Synonym Acid Blue 90, Coomassieg250, CBBG
Solubility Information Soluble in hot water (50mg/mL), ethanol (40mg/mL), methanol and 10% trichloroacetic acid. Soluble in water (1mg/mL) yielding dark Blue Solution.
InChI Key CMGWKYXJIYYESN-UHFFFAOYSA-M
Molecular Weight (g/mol) 854.02
Loss on Drying 0.045
UV_Vis λ max (methanol) 608 ± 3 nm
Color Dark Reddish Purple
Quantity 100 g
Physical Form Powder
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.