Specifications
| For Use With (Application) | There is no risk of excessive staining. Instead, superior visibility for each band is achieved when the gel is submerged overnight. Carefully mix COOMASSIE nano up and down few times before use. If the gel is thicker than mini-gel (1mm), it will take longer to incubate. The staining solution may contain blue clumps. This is normal and easily dissolved in distilled water |
| Quantity | 500 mL |
| Product Line | Novus |