Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COOMBS Polyclonal Antibody (Z-1)
GREENER_CHOICE

Catalog No. 01670412 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-412 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-412 Supplier Invitrogen™ Supplier No. PA17678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-7678 detects COOMBS protein in human samples. PA1-7678 has been successfully used in agglutination applications. The PA1-7678 immunogen is a COOMBS protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen COOMBS
Applications Agglutination
Classification Polyclonal
Clone Z-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen COOMBS.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.