Learn More
CoorsTek™ Porcelain Büchner Funnels with Double Wall, Fixed Perforated Plate
Specifications
Material Porcelain
For Use With (Application) Designed for circulation of water or steam
Includes Fixed perforated plate, inlet and outlet tubulatures, double wall
Quantity Each
Product Type Buchner Funnel
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Plate Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Plate Height (Metric) Price Quantity & Availability  
10-354D
View Documents
CoorsTek™
60270
500 mL 11cm 21.7 cm N/A N/A N/A
10-354F
View Documents
CoorsTek™
60273
2000 mL 18.6cm 27 cm N/A N/A N/A
Description
Description
For circulation of water or steam. Inlet and outlet tubulatures. Glazed inside and out except for rim.
Specifications
Specifications
Porcelain
Fixed perforated plate, inlet and outlet tubulatures, double wall
Buchner Funnel
Designed for circulation of water or steam
Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.