Learn More
CoorsTek™ Discs
Specifications
Specifications
| Material | High Alumina |
| For Use With (Application) | Ideal for high-temperature work demanding contamination-free results |
| Thickness (Metric) | 5 mm |
| Diameter (English) | 5.98 in. |
| Diameter (Metric) | 152 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.