Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568382 Supplier Invitrogen™ Supplier No. PA568382
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COP1 Polyclonal Antibody for Western Blot, IHC (P)

Caspase inhibitor. Acts as a regulator of procaspase-1/CASP1 activation implicated in the regulation of the proteolytic maturation of pro-interleukin-1 beta (IL1B) and its release during inflammation. Inhibits the release of IL1B in response to LPS in monocytes. Also induces NF-kappa-B activation during the pro-inflammatory cytokine response. Also able to inhibit CASP1-mediated neuronal cell death, TNF-alpha, hypoxia-, UV-, and staurosporine-mediated cell death but not ER stress-mediated cell death. Acts by preventing activation of caspases CASP1 and CASP4, possibly by preventing the interaction between CASP1 and RIPK2. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CARD16
Gene Accession No. Q5EG05
Gene Alias CARD only domain-containing protein 1; CARD only protein; CARD16; CARD-only protein 1; caspase recruitment domain family member 16; caspase recruitment domain family, member 16; caspase recruitment domain-containing protein 16; Caspase recruitment domain-only protein 1; caspase-1 dominant-negative inhibitor pseudo-ICE; caspase-1 inhibitor COP; Constitutive photomorphogenesis protein 1 homolog; constitutive photomorphogenic protein 1; COP; COP1; E3 ubiquitin-protein ligase COP1; E3 ubiquitin-protein ligase RFWD2; hCOP1; Pseudo interleukin-1 beta converting enzyme; pseudo interleukin-1beta converting enzyme; PSEUDO-ICE; pseudo-IL1B-converting enzyme; putative ubiquitin ligase COP1; RFWD2; ring finger and WD repeat domain 2; ring finger and WD repeat domain 2, E3 ubiquitin protein ligase; RING finger and WD repeat domain protein 2; RING finger protein 200; RING-type E3 ubiquitin transferase RFWD2; RNF200
Gene Symbols CARD16
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the N terminal of human CARD16.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114769
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.