Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141993 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141993 Supplier Invitrogen™ Supplier No. PA5141993
  • $508.00
    $466.65 / Each of 1

    Save $41.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 2 (NP_443121.1) only. In addition there may be cross-reaction with INCA (GeneID 440068; NP_001007233.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.

Caspase inhibitor. Acts as a regulator of procaspase-1/CASP1 activation implicated in the regulation of the proteolytic maturation of pro-interleukin-1 beta (IL1B) and its release during inflammation. Inhibits the release of IL1B in response to LPS in monocytes. Also induces NF-kappa-B activation during the pro-inflammatory cytokine response. Also able to inhibit CASP1-mediated neuronal cell death, TNF-alpha, hypoxia-, UV-, and staurosporine-mediated cell death but not ER stress-mediated cell death. Acts by preventing activation of caspases CASP1 and CASP4, possibly by preventing the interaction between CASP1 and RIPK2. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COP1
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CARD16
Gene Accession No. Q5EG05
Gene Alias ARABIDOPSIS THALIANA CONSTITUTIVE PHOTOMORPHOGENIC 1; ATCOP1; CARD only domain-containing protein 1; CARD only protein; CARD16; CARD-only protein 1; caspase recruitment domain family member 16; caspase recruitment domain family, member 16; caspase recruitment domain-containing protein 16; Caspase recruitment domain-only protein 1; caspase-1 dominant-negative inhibitor pseudo-ICE; caspase-1 inhibitor COP; Constitutive photomorphogenesis protein 1 homolog; CONSTITUTIVE PHOTOMORPHOGENIC 1; constitutive photomorphogenic protein 1; COP; COP1; DEETIOLATED MUTANT 340; DET340; E3 ubiquitin-protein ligase COP1; E3 ubiquitin-protein ligase RFWD2; EMB168; EMBRYO DEFECTIVE 168; FUS1; FUSCA 1; hCOP1; Pseudo interleukin-1 beta converting enzyme; pseudo interleukin-1beta converting enzyme; PSEUDO-ICE; pseudo-IL1B-converting enzyme; putative ubiquitin ligase COP1; RFWD2; ring finger and WD repeat domain 2; ring finger and WD repeat domain 2, E3 ubiquitin protein ligase; RING finger and WD repeat domain protein 2; RING finger protein 200; RING-type E3 ubiquitin transferase RFWD2; RNF200; T21L14.11; T21L14_11
Gene Symbols CARD16
Host Species Goat
Immunogen Peptide with sequence C-ETLGLSAGPIPGN.
Purification Method Antigen affinity chromatography, Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114769
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.