Learn More
Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack
Shop All Copan Diagnostics Inc. Products
Description
Calibrated 1 and 10μL plastic disposable inoculating loops and needles
Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.
- Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
- Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
- Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
- Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
- Color coded for easy product identification
- Packed in tamper-proof bags to guarantee sterility
- Certificate of calibration included in each case
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Disposable | Single-use |
| Volume | 1 μL |
| Sterility | Sterile |
| Material | Plastic |
| Color | Dark Green |
| Product Type | Inoculating Loop, Rigid |
| Includes | Evans blue dye certificate of calibration |
| Packaging Type | Tamperproof Ziplock resealable bag |
| Calibrated | Yes |