Learn More
Copan Diagnostics ESwab™Patented Sample Collection and Delivery System
GSA_VASDP
Catalog No. 23600901 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Swab Configuration:
Minitip flocked
Regular flocked
Single Applicator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Swab Configuration
23-600-901 Minitip flocked
23-600-900 Regular flocked
23-600-902 Single Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-600-901 Supplier Copan Diagnostics Supplier No. 481C
Add to Cart
Add to Cart
This item is not returnable. View return policy
New liquid-based transport for microbiology
  • Patented process sprays tips with short strands of nylon fiber that bond in a perpendicular fashion
  • Strong capillary action between the fibers on the flocked tip keeps the sample very close to the surface
  • Sample rapidly and completely elutes from the flocked swab as soon as it is placed in the transport medium
  • Various molded tips correct for different parts of the human anatomy
  • Provides 1mL of sample suspension for Gram stain and multiple culture analysis
  • Maintains viability of aerobes, anaerobes and fastidious bacteria for up to 48 hours
  • Suitable platform for automated liquid handling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Specimen Collection Kit
Sterility Sterile
Swab Configuration Minitip flocked
For Use With (Application) Maintains viability of aerobes, anaerobes and fastidious Bacteria for up to 48 Hours
Includes Polypropylene screw cap tube containing 1mL of liquid Amies medium, minitip swab with flocked Nylon Fiber tip
Quantity 50 Pk.
Tip Type Minitip Flocked