Learn More
Description
Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.
- Ergonomic and Anatomic Design: Improves patient comfort and specimen collection
- Rapid Automatic Elution: Instant and spontaneous release of sample into liquid media
- Measurable and Consistent Uptake and Transfer: Quantitative measurement and improved test sensitivity
- Patient Friendly: Anatomical design and a soft texture for rapidly and efficiently dislodging cells
- Compatible with Multiple Applications: Rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, bacteriology and virology culture
- Safe and Convenient: Molded breakpoint
Specifications
Specifications
| Tip Type | Flexible Minitip |
| Applicator Material | Plastic |
| Tip Material | Nylon |
| Sterility | Sterile |
| Packaging | Individually-wrapped in peel pouch |
| Length (Metric) Shaft | 100 mm |
| Length (English) Shaft | 3.94 in. |
| Product Type | Specimen Collection Swab |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
