Learn More
Description
Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.
- Ergonomic and Anatomic Design: Improves patient comfort and specimen collection
- Rapid Automatic Elution: Instant and spontaneous release of sample into liquid media
- Measurable and Consistent Uptake and Transfer: Quantitative measurement and improved test sensitivity
- Patient Friendly: Anatomical design and a soft texture for rapidly and efficiently dislodging cells
- Compatible with Multiple Applications: Rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, bacteriology and virology culture
- Safe and Convenient: Molded breakpoint
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Applicator Material | Plastic |
| Tip Type | Minitip flocked |
| Sterility | Sterile |
| Packaging | Individually-wrapped in peel pouch |
| Length (Metric) Shaft | 100 mm |
| Length (English) Shaft | 3.94 in. |
| Product Type | Specimen Collection Swab |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

