Learn More
Copan Diagnostics T-Shaped Spreaders
SDP
Catalog No. 23600895 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Quantity:
1/Peel Pk., 500 Spreaders/Box
10/Peel Pk., 500 Spreaders/Box
5/Peel Pk., 1000 Spreaders/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-600-895 5/Peel Pk., 1000 Spreaders/Box
23-600-894 1/Peel Pk., 500 Spreaders/Box
23-600-896 10/Peel Pk., 500 Spreaders/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-600-895 Supplier Copan Diagnostics Supplier No. COPTS5
Add to Cart
Add to Cart
This item is not returnable. View return policy
T-shaped spreaders
T-Shaped Plastic Spreaders
  • Designed for spreading and dispersing liquids across the surface of agar culture plates
  • Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
  • Smooth, rounded surfaces prevent any cutting or gouging of agar during inoculation of culture plates
  • T-Shape design means that even pressure is applied across the entire length of the T-bar in contact with the agar
  • Available individually sterile wrapped or in packs of five or ten pieces in a peel pack
Specifications
Product Type Cell Spreader
Quantity 5/Peel Pk., 1000 Spreaders/Box
For Use With (Equipment) Culture Plates
Shape T-Shape
Sterility Sterile
Material Plastic
For Use With (Application) Designed for spreading and dispersing liquids across the surface of agar culture plates, used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples