Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ESwab™

Catalog No. R723482
Change view
Click to view available options
Closure Color:
Blue
Green
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
R723482 Blue
R723481 Green
R723480 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R723482 Supplier Thermo Scientific™ Supplier No. R723482
Add to Cart
Add to Cart
This item is not returnable. View return policy

Expand testing capabilities and experience savings by replacing multiple products with ESwab™, a liquid transport swab system compatible with automated specimen processing.

Expand testing capabilities and make specimen collection and preservation simple with the ESwab™ System. ESwab is a liquid-based, multipurpose open platform, collection and preservation system that maintains viability of aerobic, anaerobic and fastidious bacteria for up to 48 hours at refrigerator and room temperature. The multipurpose ESwab offers significant cost savings—use it for automated specimen processing, gram stains, traditional bacteriology culture, rapid antigen tests* and molecular assays*.

*Self-validation required

Collect, snap and elute with ESwab; that is all it takes to simplify and streamline specimen collection.

In the testing laboratory, the ESwab cap is unscrewed and removed, and the swab applicator stick is securely attached to the cap. This allows the operator to conveniently remove the swab and perform various microbiology analyses, using the tube cap as a handle to hold and manipulate the swab.

Automated Specimen Processing
  • A collection and transport system cleared by the FDA to maintain viability of aerobes, anaerobes and fastidious bacteria for up to 48 hours at room temperature and refrigerator temperature*
  • Polypropylene screw-cap tube with internal conical shape filled with 1mL Liquid Aimes preservation medium elutes the entire sample upon contact
  • The only liquid transport swab system which allows for automated specimen processing with the COPAN WASP™ line of instruments
Cost Savings
  • Expands testing capabilities with identical aliquots of liquid sample suspension, allowing for multiple tests from the same specimen
  • Allows for significant cost savings by replacing multiple products with just one, eliminating the need to stock multiple types of swabs
  • Creates more consistent gram stains as the entire specimen elutes from the FLOQSwab™, creating a homogenous suspension for a more precise and even smear
*Product tested and validated in full compliance with CLSI M40-A: Quality Control of Microbiological Transport System standard, including Neisseria gonorrhoeae survival at 24 hours.

COPAN WASP instrumentation and accessories and Eswab collection and transport products are available through Thermo Fisher Scientific in the U.S.A. only. Please contact your local sales representative for more information.

Eswab and WASP are trademarks or registered trademarks of COPAN S.P.A. All other trademarks are property of Thermo Fisher Scientific and its subsidiaries.
TRUSTED_SUSTAINABILITY

Specifications

Product Type NPG Flocked Collection Kit
Format Tube
Swab Configuration Single Applicator
Closure Color Blue
Description ESwab NPG Flocked Collection Kit
For Use With (Application) Nasopharynx and pediatric sample collection
Quantity 50 Kits/Pk.
Shaft Material Plastic
Storage Requirements 5° to 25°C
Tip Material Nylon Fiber
Tip Type Flocked Minitip
Volume (Metric) 1 mL
Show More Show Less

Research Use Only Authorization required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.