Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPG2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7207270 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0722 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0722 Supplier Proteintech Group Inc Supplier No. 161111AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network.

Specifications

Antigen COPG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene COPG2
Gene Accession No. Q9QXK3, Q9UBF2
Gene Alias 2 COP, Coatomer subunit gamma 2, COPG2, Gamma 2 coat protein, Gamma 2 COP
Gene Symbols COPG2
Host Species Rabbit
Immunogen COPG2 Fusion Protein Ag9034
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26958, 54160
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.