Learn More
Medchemexpress LLC Copper-ATSP | 68341-12-8 | 99.4% | C18H18CuN6S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13982610MG
Encompass_Preferred
CuATSP is a copper bis(thiosemicarbazone) research compound used to inhibit ferroptotic cell death. It is reported to be roughly 20-fold more potent than the related CuATSM complex and is supplied as a solid for cellular and biochemical studies of ferroptosis.
  • Potent inhibitor of ferroptotic cell death.
  • Approximately 20-fold more potent than related analogues.
  • High purity (99.4%).
  • Solid, brown to reddish brown appearance.
  • Intended for biochemical and cellular research applications.
Catalog No. 50-002-59733