Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Gluconate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601838 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-838 125 g Poly Bottle
18-601-839 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-838 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1334125GM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

C1334, 527-09-3, C12H22CuO14

Spectrum™ Chemical Copper Gluconate, USP is used as a nutrient and dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 527-09-3
Molecular Formula C12H22CuO14
Molecular Weight (g/mol) 453.84
InChI Key OCUCCJIRFHNWBP-IYEMJOQQSA-L
IUPAC Name copper(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Cu++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 102%
Grade USP
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.