Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) Hexafluoro-2,4-pentanedionate 1,5-Cyclooctadiene Complex 95.0+%, TCI America™
SDP

Catalog No. C26101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26101G 1 g
C26105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26101G Supplier TCI America Supplier No. C26101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86233-74-1
Molecular Formula C13H14CuF6O2
Molecular Weight (g/mol) 379.79
MDL Number MFCD00156517
InChI Key WSAPIYREFIKPSH-OJKFPHAYSA-N
Synonym copper i hexafluoro-2,4-pentanedionate-cyclooctadiene complex, copper i hexafluoroacetylacetonate cyclooctadiene complex
PubChem CID 92043567
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene (3Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one copper
SMILES [Cu].C1C\C=C/CC\C=C/1.O\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 378.78
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Copper(I) Hexafluoro-2,4-pentanedionate 1,5-Cyclooctadiene Complex
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.