Learn More
Copper(I) oxide, 99% (metals basis)
Description
Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Purple-Red |
| Physical Form | Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% (Metal basis) |
| Formula Weight | 143.08 |
| Odor | Odorless |
| Appearance | Purple-red powder |
| Chemical Name or Material | Copper(I) oxide |
Safety and Handling
| Hazard Category | H400-H410-H302: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. |
| DOTInformation | DOT Class: 9, Packing Group: III |
| EINECSNumber | 215-270-7 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.